Assay Details
| Gene Symbol: LOC4325777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765851.2 | XP_015621337.1 | 6 - 7 | 710 | 79 | |
| GenBank mRNA | AK062011.1 | - | 2 - 3 | 166 | 79 | |
| AK070568.1 | - | 6 - 7 | 679 | 79 | ||
| AK099208.1 | - | 6 - 7 | 646 | 79 | ||
More Information
Set Membership: |
|

LOC4325777
Genomic Map