Assay Details
| Gene Symbol: LOC4325320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765975.2 | XP_015621461.1 | 1 - 2 | 4165 | 66 | |
| GenBank mRNA | AJ620677.1 | - | 1 - 2 | 202 | 66 | |
| AK065375.1 | - | 1 - 2 | 2344 | 66 | ||
More Information
Set Membership: |
|

LOC4325320
Genomic Map