Assay Details
| Gene Symbol: LOC4325320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765975.2 | XP_015621461.1 | 3 - 4 | 4387 | 57 | |
| GenBank mRNA | AJ620677.1 | - | 3 - 4 | 424 | 57 | |
| AK065375.1 | - | 3 - 4 | 2566 | 57 | ||
More Information
Set Membership: |
|

LOC4325320
Genomic Map