Assay Details
| Gene Symbol: LOC4326689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771572.2 | XP_015627058.1 | 1 | 462 | 90 | |
| GenBank mRNA | AK122071.1 | - | 1 | 452 | 90 | |
More Information
Set Membership: |
|

LOC4326689
Genomic Map