Assay Details
Gene Symbol: LOC4326574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766584.1 | XP_015622070.1 | 22 - 23 | 2624 | 115 | |
GenBank mRNA | AK099822.1 | - | 22 - 23 | 2618 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
