Assay Details
Gene Symbol: LOC4326574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766584.1 | XP_015622070.1 | 5 - 6 | 527 | 107 | |
XM_015766588.2 | XP_015622074.1 | 4 - 5 | 316 | 107 | ||
GenBank mRNA | AK099822.1 | - | 5 - 6 | 521 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
