Assay Details
| Gene Symbol: LOC4326553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772783.2 | XP_015628269.1 | 1 - 2 | 213 | 104 | |
| XM_015772793.2 | XP_015628279.1 | 1 - 2 | 213 | 104 | ||
| XM_015772802.2 | XP_015628288.1 | 1 - 2 | 213 | 104 | ||
| GenBank mRNA | AK065688.1 | - | 1 - 2 | 219 | 104 | |
More Information
Set Membership: |
|

LOC4326553
Genomic Map