Assay Details
Gene Symbol: LOC4326935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765654.2 | XP_015621140.1 | 1 - 2 | 577 | 61 | |
GenBank mRNA | AK063691.1 | - | 2 - 3 | 341 | 61 | |
AY333185.1 | - | 1 - 2 | 549 | 61 | ||
X57327.1 | - | 1 - 2 | 520 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
