Assay Details
| Gene Symbol: LOC4326167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795699.2 | XP_015651185.1 | 19 - 20 | 2879 | 85 | |
| GenBank mRNA | AK058804.1 | - | 5 - 6 | 816 | 85 | |
| AK101026.1 | - | 19 - 20 | 2845 | 85 | ||
More Information
Set Membership: |
|

LOC4326167
Genomic Map