Assay Details
| Gene Symbol: LOC4326167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795699.2 | XP_015651185.1 | 1 - 2 | 281 | 69 | |
| GenBank mRNA | AK101026.1 | - | 1 - 2 | 247 | 69 | |
More Information
Set Membership: |
|

LOC4326167
Genomic Map