Assay Details
| Gene Symbol: LOC4326167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795699.2 | XP_015651185.1 | 9 - 10 | 1133 | 90 | |
| GenBank mRNA | AK101026.1 | - | 9 - 10 | 1099 | 90 | |
More Information
Set Membership: |
|

LOC4326167
Genomic Map