Assay Details
| Gene Symbol: LOC4324792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765582.2 | 12 - 13 | 1138 | 65 | ||
| XM_015765583.2 | XP_015621069.1 | 12 - 13 | 922 | 65 | ||
More Information
Set Membership: |
|

LOC4324792
Genomic Map