Assay Details
| Gene Symbol: LOC4325081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766060.2 | XP_015621546.1 | 3 - 4 | 2057 | 73 | |
| GenBank mRNA | AK065101.1 | - | 4 - 5 | 1866 | 73 | |
| AK072910.1 | - | 3 - 4 | 1850 | 73 | ||
| AK073144.1 | - | 3 - 4 | 1890 | 73 | ||
| AK100486.1 | - | 2 - 3 | 1304 | 73 | ||
| AK121648.1 | - | 3 - 4 | 1893 | 73 | ||
More Information
Set Membership: |
|

LOC4325081
Genomic Map