Assay Details
| Gene Symbol: LOC4324993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756665.2 | XP_015612151.1 | 1 - 2 | 939 | 102 | |
| GenBank mRNA | AK102849.1 | - | 1 - 2 | 841 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756665.2 transcripts.
Set Membership: |
|

LOC4324993
Genomic Map