Assay Details
| Gene Symbol: LOC9269568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771401.2 | XP_015626887.1 | 1 - 2 | 603 | 72 | |
| GenBank mRNA | AB239918.1 | - | 1 - 2 | 341 | 72 | |
| AK242651.1 | - | 1 - 2 | 440 | 72 | ||
More Information
Set Membership: |
|

LOC9269568
Genomic Map