Assay Details
| Gene Symbol: LOC9269568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771401.2 | XP_015626887.1 | 6 - 7 | 1084 | 68 | |
| GenBank mRNA | AB239918.1 | - | 6 - 7 | 822 | 68 | |
| AK242651.1 | - | 6 - 7 | 921 | 68 | ||
More Information
Set Membership: |
|

LOC9269568
Genomic Map