Assay Details
| Gene Symbol: LOC4325107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773549.2 | XP_015629035.1 | 4 | 3318 | 157 | |
| GenBank mRNA | AK061290.1 | - | 1 | 852 | 157 | |
More Information
Set Membership: |
|

LOC4325107
Genomic Map