Assay Details
Gene Symbol: LOC4325020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765404.1 | XP_015620890.1 | 10 - 11 | 2219 | 153 | |
GenBank mRNA | AK103514.1 | - | 10 - 11 | 2225 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
