Assay Details
| Gene Symbol: LOC9271802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793086.2 | XP_015648572.1 | 5 - 6 | 1706 | 89 | |
| GenBank mRNA | AK064782.1 | - | 5 - 6 | 1136 | 89 | |
| AK068197.1 | - | 104 | 89 | |||
More Information
Set Membership: |
|

LOC9271802
Genomic Map