Assay Details
| Gene Symbol: LOC9271802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793086.2 | XP_015648572.1 | 7 - 8 | 1883 | 149 | |
| GenBank mRNA | AK064782.1 | - | 7 - 8 | 1313 | 149 | |
| AK068197.1 | - | 281 | 149 | |||
More Information
Set Membership: |
|

LOC9271802
Genomic Map