Assay Details
| Gene Symbol: LOC9271802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793086.2 | XP_015648572.1 | 9 - 10 | 2340 | 103 | |
| GenBank mRNA | AK062123.1 | - | 3 - 4 | 516 | 103 | |
| AK064782.1 | - | 8 - 9 | 3033 | 103 | ||
| AK068197.1 | - | 2002 | 103 | |||
More Information
Set Membership: |
|

LOC9271802
Genomic Map