Assay Details
| Gene Symbol: LOC4327741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762492.2 | XP_015617978.1 | 3 - 4 | 475 | 80 | |
| GenBank mRNA | AK120182.1 | - | 2 - 3 | 160 | 80 | |
| BT129172.1 | - | 2 - 3 | 154 | 80 | ||
| BT130740.1 | - | 2 - 3 | 158 | 80 | ||
More Information
Set Membership: |
|

LOC4327741
Genomic Map