Assay Details
| Gene Symbol: LOC4327741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762428.2 | XP_015617914.1 | 5 - 6 | 646 | 114 | |
| XM_015762492.2 | XP_015617978.1 | 4 - 5 | 570 | 114 | ||
| GenBank mRNA | AK120182.1 | - | 3 - 4 | 255 | 114 | |
| BT129172.1 | - | 3 - 4 | 249 | 114 | ||
| BT130740.1 | - | 3 - 4 | 253 | 114 | ||
| BT131555.1 | - | 4 - 5 | 397 | 114 | ||
More Information
Set Membership: |
|

LOC4327741
Genomic Map