Assay Details
| Gene Symbol: LOC4325761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766829.2 | XP_015622315.1 | 4 - 5 | 1311 | 119 | |
| GenBank mRNA | AK101395.1 | - | 4 - 5 | 486 | 119 | |
More Information
Set Membership: |
|

LOC4325761
Genomic Map