Assay Details
| Gene Symbol: LOC4325761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766829.2 | XP_015622315.1 | 6 - 7 | 1547 | 100 | |
| GenBank mRNA | AK061271.1 | - | 5 - 6 | 806 | 100 | |
| AK101395.1 | - | 6 - 7 | 722 | 100 | ||
More Information
Set Membership: |
|

LOC4325761
Genomic Map