Assay Details
| Gene Symbol: LOC4325761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766829.2 | XP_015622315.1 | 7 - 8 | 1594 | 131 | |
| GenBank mRNA | AK061271.1 | - | 6 - 7 | 853 | 131 | |
| AK101395.1 | - | 7 - 8 | 769 | 131 | ||
More Information
Set Membership: |
|

LOC4325761
Genomic Map