Assay Details
| Gene Symbol: LOC4327993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771048.2 | XP_015626534.1 | 4 - 5 | 637 | 88 | |
| GenBank mRNA | AK070603.1 | - | 4 - 5 | 590 | 88 | |
| AK101537.1 | - | 4 - 5 | 569 | 88 | ||
| GU120333.1 | - | 4 - 5 | 590 | 88 | ||
More Information
Set Membership: |
|

LOC4327993
Genomic Map