Assay Details
| Gene Symbol: LOC4328790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770828.2 | XP_015626314.1 | 10 - 11 | 2541 | 115 | |
| GenBank mRNA | AK103143.1 | - | 10 - 11 | 2551 | 115 | |
More Information
Set Membership: |
|

LOC4328790
Genomic Map