Assay Details
| Gene Symbol: LOC4328790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770828.2 | XP_015626314.1 | 11 - 12 | 2586 | 80 | |
| XM_015770829.2 | XP_015626315.1 | 10 - 11 | 2688 | 80 | ||
| GenBank mRNA | AK103143.1 | - | 11 - 12 | 2596 | 80 | |
More Information
Set Membership: |
|

LOC4328790
Genomic Map