Assay Details
| Gene Symbol: LOC4328790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770828.2 | XP_015626314.1 | 2 - 3 | 678 | 104 | |
| XM_015770829.2 | XP_015626315.1 | 2 - 3 | 672 | 104 | ||
| GenBank mRNA | AK103143.1 | - | 2 - 3 | 680 | 104 | |
More Information
Set Membership: |
|

LOC4328790
Genomic Map