Assay Details
| Gene Symbol: LOC4328790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770828.2 | XP_015626314.1 | 3 - 4 | 790 | 84 | |
| XM_015770829.2 | XP_015626315.1 | 3 - 4 | 784 | 84 | ||
More Information
Set Membership: |
|

LOC4328790
Genomic Map