Assay Details
| Gene Symbol: LOC4328790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770828.2 | XP_015626314.1 | 5 - 6 | 1018 | 103 | |
| XM_015770829.2 | XP_015626315.1 | 5 - 6 | 1012 | 103 | ||
| GenBank mRNA | AK103143.1 | - | 5 - 6 | 1020 | 103 | |
More Information
Set Membership: |
|

LOC4328790
Genomic Map