Assay Details
| Gene Symbol: LOC4328790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770828.2 | XP_015626314.1 | 6 - 7 | 1132 | 73 | |
| XM_015770829.2 | XP_015626315.1 | 6 - 7 | 1126 | 73 | ||
| GenBank mRNA | AK103143.1 | - | 6 - 7 | 1134 | 73 | |
More Information
Set Membership: |
|

LOC4328790
Genomic Map