Assay Details
| Gene Symbol: LOC4349332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757479.2 | XP_015612965.1 | 11 - 12 | 1016 | 72 | |
| GenBank mRNA | AK068963.1 | - | 11 - 12 | 1015 | 72 | |
| AK099250.1 | - | 11 - 12 | 999 | 72 | ||
More Information
Set Membership: |
|

LOC4349332
Genomic Map