Assay Details
| Gene Symbol: LOC4349332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757479.2 | XP_015612965.1 | 1 - 2 | 194 | 126 | |
| GenBank mRNA | AK068963.1 | - | 1 - 2 | 193 | 126 | |
| AK099250.1 | - | 1 - 2 | 177 | 126 | ||
More Information
Set Membership: |
|

LOC4349332
Genomic Map