Assay Details
| Gene Symbol: LOC4349332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757479.2 | XP_015612965.1 | 6 - 7 | 566 | 76 | |
| GenBank mRNA | AK068963.1 | - | 6 - 7 | 565 | 76 | |
| AK099250.1 | - | 6 - 7 | 549 | 76 | ||
More Information
Set Membership: |
|

LOC4349332
Genomic Map