Assay Details
| Gene Symbol: LOC4349332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757479.2 | XP_015612965.1 | 8 - 9 | 742 | 65 | |
| GenBank mRNA | AK068963.1 | - | 8 - 9 | 741 | 65 | |
| AK099250.1 | - | 8 - 9 | 725 | 65 | ||
More Information
Set Membership: |
|

LOC4349332
Genomic Map