Assay Details
| Gene Symbol: LOC4349344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758536.2 | XP_015614022.1 | 1 - 2 | 430 | 107 | |
| GenBank mRNA | AK068935.1 | - | 1 - 2 | 307 | 107 | |
More Information
Set Membership: |
|

LOC4349344
Genomic Map