Assay Details
| Gene Symbol: LOC4351586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765080.2 | XP_015620566.1 | 9 - 10 | 1243 | 104 | |
| XM_015765082.2 | XP_015620568.1 | 9 - 10 | 1017 | 104 | ||
| GenBank mRNA | AK243345.1 | - | 9 - 10 | 1247 | 104 | |
More Information
Set Membership: |
|

LOC4351586
Genomic Map