Assay Details
| Gene Symbol: LOC4351779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763128.1 | XP_015618614.1 | 11 - 12 | 916 | 77 | |
| GenBank mRNA | AK072548.1 | - | 10 - 11 | 926 | 77 | |
| AY224480.1 | - | 11 - 12 | 864 | 77 | ||
More Information
Set Membership: |
|

LOC4351779
Genomic Map