Assay Details
| Gene Symbol: LOC4351779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763128.1 | XP_015618614.1 | 13 - 14 | 1022 | 78 | |
| GenBank mRNA | AK072548.1 | - | 12 - 13 | 1032 | 78 | |
| AY224480.1 | - | 13 - 14 | 970 | 78 | ||
More Information
Set Membership: |
|

LOC4351779
Genomic Map