Assay Details
| Gene Symbol: LOC4351779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763128.1 | XP_015618614.1 | 1 - 2 | 162 | 92 | |
| GenBank mRNA | AY224480.1 | - | 1 - 2 | 110 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015763128.1 transcripts.
Set Membership: |
|

LOC4351779
Genomic Map