Assay Details
| Gene Symbol: LOC4351779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763128.1 | XP_015618614.1 | 6 - 7 | 561 | 120 | |
| GenBank mRNA | AK072548.1 | - | 5 - 6 | 571 | 120 | |
| AY224480.1 | - | 6 - 7 | 509 | 120 | ||
More Information
Set Membership: |
|

LOC4351779
Genomic Map