Assay Details
| Gene Symbol: LOC4351779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763128.1 | XP_015618614.1 | 8 - 9 | 717 | 105 | |
| GenBank mRNA | AK072548.1 | - | 7 - 8 | 727 | 105 | |
| AY224480.1 | - | 8 - 9 | 665 | 105 | ||
More Information
Set Membership: |
|

LOC4351779
Genomic Map