Assay Details
| Gene Symbol: LOC4351779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763128.1 | XP_015618614.1 | 9 - 10 | 776 | 101 | |
| GenBank mRNA | AK072548.1 | - | 8 - 9 | 786 | 101 | |
| AY224480.1 | - | 9 - 10 | 724 | 101 | ||
More Information
Set Membership: |
|

LOC4351779
Genomic Map