Assay Details
| Gene Symbol: LOC4352110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764728.2 | XP_015620214.1 | 12 - 13 | 1749 | 92 | |
| GenBank mRNA | AK065546.1 | - | 13 - 14 | 1726 | 92 | |
More Information
Set Membership: |
|

LOC4352110
Genomic Map