Assay Details
| Gene Symbol: LOC9267937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763043.2 | XP_015618529.1 | 2 | 915 | 95 | |
| GenBank mRNA | AK063578.1 | - | 1 | 345 | 95 | |
More Information
Set Membership: |
|

LOC9267937
Genomic Map