Assay Details
| Gene Symbol: LOC4352268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764003.2 | XP_015619489.1 | 1 - 2 | 656 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352268| Gene Symbol: LOC4352268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764003.2 | XP_015619489.1 | 1 - 2 | 656 | 85 | |
Set Membership: |
|