Assay Details
| Gene Symbol: LOC4352780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765332.1 | XP_015620818.1 | 1 - 2 | 299 | 89 | |
| GenBank mRNA | AK100618.1 | - | 1 - 2 | 300 | 89 | |
| FJ940211.1 | - | 1 - 2 | 53 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765332.1 transcripts.
Set Membership: |
|

LOC4352780
Genomic Map