Assay Details
| Gene Symbol: LOC4352780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765331.2 | XP_015620817.1 | 6 - 7 | 1227 | 76 | |
| XM_015765332.1 | XP_015620818.1 | 6 - 7 | 1226 | 76 | ||
| GenBank mRNA | AB118014.1 | - | 5 - 6 | 864 | 76 | |
| AK100618.1 | - | 6 - 7 | 1227 | 76 | ||
| AK105660.1 | - | 6 - 7 | 1227 | 76 | ||
| FJ940211.1 | - | 6 - 7 | 980 | 76 | ||
More Information
Set Membership: |
|

LOC4352780
Genomic Map